Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
MRPL13 Antibody (OTI6A11), Alexa Fluor™ 594, Novus Biologicals™

Mouse Monoclonal Antibody
Supplier: Novus Biologicals NBP272780AF594
Description
MRPL13 Monoclonal specifically detects MRPL13 in Human, Mouse, Rat samples. It is validated for Western Blot.Specifications
MRPL13 | |
Monoclonal | |
Alexa Fluor 594 | |
50 mM sodium borate with 0.05% sodium azide | |
MRPL13 | |
Full length recombinant protein of human MRPL13 (NP_054797) produced in E.coli. | |
0.1 mL | |
Primary | |
Human, Mouse, Rat | |
IgG2b |
Western Blot | |
OTI6A11 | |
Western Blot | |
39S ribosomal protein L13, mitochondrial, L13, L13A, L13mtRPML13, mitochondrial ribosomal protein L13, MRP-L13, RPL13 | |
Mouse | |
Affinity Purified | |
Core ESC Like Genes, Stem Cell Markers | |
28998.0 | |
Store at 4°C in the dark. |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction