Learn More
MRPL13 Polyclonal Antibody, Invitrogen™
Rabbit Polyclonal Antibody
Supplier: Thermo Scientific PA589633
Description
Immunogen sequence: MSSFSRAPQQ WATFARIWYL LDGKMQPPGK LAAMASIRLQ GLHKPVYHAL SDCGDHVVIM NTRHIAFSGN KWEQKVYSSH TGYPGGFRQV TAAQLHLRDP VAIVKLAIYG MLPKNLHRRT MMERLHLFPD EYIPEDILKN LVEELPQPRK IPKRLDEYTQ EEIDAFPRLW TPPEDYRL; Positive Samples: SH-SY5Y, HL-60, HT-29, A-549, HeLa, Mouse kidney, Mouse heart; Cellular Location: Mitochondrion
Mammalian mitochondrial ribosomal proteins are encoded by nuclear genes and help in protein synthesis within the mitochondrion. Mitochondrial ribosomes (mitoribosomes) consist of a small 28S subunit and a large 39S subunit. They have an estimated 75% protein to rRNA composition compared to prokaryotic ribosomes, where this ratio is reversed. Another difference between mammalian mitoribosomes and prokaryotic ribosomes is that the latter contain a 5S rRNA. Among different species, the proteins comprising the mitoribosome differ greatly in sequence, and sometimes in biochemical properties, which prevents easy recognition by sequence homology. This gene encodes a 39S subunit protein.
Specifications
MRPL13 | |
Polyclonal | |
Unconjugated | |
MRPL13 | |
1110002D09Rik, 39S ribosomal protein L13, mitochondrial, L13, L13A, L13mt, Mitochondrial large ribosomal subunit protein uL13m, mitochondrial ribosomal protein L13, Mrpl13, MRP-L13, RPL13, RPML13 | |
Rabbit | |
Affinity Chromatography | |
RUO | |
28998, 299938, 68537 | |
-20° C, Avoid Freeze/Thaw Cycles | |
Liquid |
Immunohistochemistry (Paraffin), Western Blot | |
0.19 mg/mL | |
PBS with 50% glycerol and 0.02% sodium azide, pH 7.3 | |
Q9BYD1, Q9D1P0 | |
MRPL13 | |
Recombinant fusion protein containing a sequence corresponding to amino acids 1-178 of human MRPL13 (NP_054797.2). | |
100 μL | |
Primary | |
Human, Mouse, Rat | |
Antibody | |
IgG |
Your input is important to us. Please complete this form to provide feedback related to the content on this product.