Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ MRPL14 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PA551008
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PA551008 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PA551008 Supplier Invitrogen™ Supplier No. PA551008
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

MRPL14 Polyclonal Antibody detects endogenous levels of MRPL14 protein.

Mammalian mitochondrial ribosomal proteins are encoded by nuclear genes and help in protein synthesis within the mitochondrion. Mitochondrial ribosomes (mitoribosomes) consist of a small 28S subunit and a large 39S subunit. They have an estimated 75% protein to rRNA composition compared to prokaryotic ribosomes, where this ratio is reversed. Another difference between mammalian mitoribosomes and prokaryotic ribosomes is that the latter contain a 5S rRNA. Among different species, the proteins comprising the mitoribosome differ greatly in sequence, and sometimes in biochemical properties, which prevents easy recognition by sequence homology. This gene encodes a 39S subunit protein. A pseudogene corresponding to this gene is found at 17p13.3.
TRUSTED_SUSTAINABILITY

Specifications

Antigen MRPL14
Applications Immunohistochemistry, Western Blot
Classification Polyclonal
Concentration 1 mg/mL
Conjugate Unconjugated
Formulation PBS with 0.5% BSA, 50% glycerol and 0.02% sodium azide; pH 7.4
Gene Mrpl14
Gene Accession No. Q6P1L8, Q7M0E7, Q9D1I6
Gene Alias 1110006I11Rik; 39S ribosomal protein L14, mitochondrial; 39S ribosomal protein L32, mitochondrial; AI846816; L14mt; L32mt; Mitochondrial large ribosomal subunit protein uL14m; mitochondrial ribosomal protein L14; mitochondrial ribosomal protein L32; Mrpl14; MRP-L14; MRPL32; MRP-L32; RMPL32; Rpml32
Gene Symbols Mrpl14
Host Species Rabbit
Immunogen Synthesized peptide derived from the C-terminal region of human MRP-L14.
Purification Method Antigen affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 301250, 64928, 68463
Target Species Human, Mouse, Rat
Content And Storage -20°C, Avoid Freeze/Thaw Cycles
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.