Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Invitrogen™ MRPL14 Polyclonal Antibody

Rabbit Polyclonal Antibody
Supplier: Invitrogen™ PA585010
Description
Immunogen sequence: FSTTGSLSAI QKMTRVRVVD NSALGNSPYH RAPRCIHVYK KNGVGKVGDQ ILLAIK.
Mammalian mitochondrial ribosomal proteins are encoded by nuclear genes and help in protein synthesis within the mitochondrion. Mitochondrial ribosomes (mitoribosomes) consist of a small 28S subunit and a large 39S subunit. They have an estimated 75% protein to rRNA composition compared to prokaryotic ribosomes, where this ratio is reversed. Another difference between mammalian mitoribosomes and prokaryotic ribosomes is that the latter contain a 5S rRNA. Among different species, the proteins comprising the mitoribosome differ greatly in sequence, and sometimes in biochemical properties, which prevents easy recognition by sequence homology. This gene encodes a 39S subunit protein. A pseudogene corresponding to this gene is found at 17p13.3.
Specifications
MRPL14 | |
Polyclonal | |
Unconjugated | |
Mrpl14 | |
1110006I11Rik; 39S ribosomal protein L14, mitochondrial; 39S ribosomal protein L32, mitochondrial; AI846816; L14mt; L32mt; Mitochondrial large ribosomal subunit protein uL14m; mitochondrial ribosomal protein L14; mitochondrial ribosomal protein L32; Mrpl14; MRP-L14; MRPL32; MRP-L32; RMPL32; Rpml32 | |
Rabbit | |
Antigen affinity chromatography | |
RUO | |
64928 | |
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
Liquid |
Immunocytochemistry, Immunofluorescence | |
0.1 mg/mL | |
PBS with 40% glycerol and 0.02% sodium azide; pH 7.2 | |
Q6P1L8 | |
Mrpl14 | |
Recombinant protein corresponding to Human MRPL14. Recombinant protein control fragment (Product #RP-108098). | |
100 μL | |
Primary | |
Human | |
Antibody | |
IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction