Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Invitrogen™ MRPL14 Polyclonal Antibody

Rabbit Polyclonal Antibody
Supplier: Invitrogen™ PA5106572
Description
Antibody detects endogenous levels of total MRPL14.
Mammalian mitochondrial ribosomal proteins are encoded by nuclear genes and help in protein synthesis within the mitochondrion. Mitochondrial ribosomes (mitoribosomes) consist of a small 28S subunit and a large 39S subunit. They have an estimated 75% protein to rRNA composition compared to prokaryotic ribosomes, where this ratio is reversed. Another difference between mammalian mitoribosomes and prokaryotic ribosomes is that the latter contain a 5S rRNA. Among different species, the proteins comprising the mitoribosome differ greatly in sequence, and sometimes in biochemical properties, which prevents easy recognition by sequence homology. This gene encodes a 39S subunit protein. A pseudogene corresponding to this gene is found at 17p13.3.
Specifications
MRPL14 | |
Polyclonal | |
Unconjugated | |
Mrpl14 | |
1110006I11Rik; 39S ribosomal protein L14, mitochondrial; 39S ribosomal protein L32, mitochondrial; AI846816; L14mt; L32mt; Mitochondrial large ribosomal subunit protein uL14m; mitochondrial ribosomal protein L14; mitochondrial ribosomal protein L32; Mrpl14; MRP-L14; MRPL32; MRP-L32; RMPL32; Rpml32 | |
Rabbit | |
Affinity chromatography | |
RUO | |
301250, 64928, 68463 | |
-20°C | |
Liquid |
Western Blot, Immunocytochemistry | |
1 mg/mL | |
PBS with 50% glycerol and 0.02% sodium azide; pH 7.4 | |
Q6P1L8, Q7M0E7, Q9D1I6 | |
Mrpl14 | |
A synthesized peptide derived from human MRPL14(Accession Q6P1L8), corresponding to amino acid residues N103-V145. | |
100 μL | |
Primary | |
Human, Mouse, Rat | |
Antibody | |
IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction