Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
MRPL15 Antibody (OTI7A3), FITC, Novus Biologicals™

Mouse Monoclonal Antibody
Supplier: Novus Biologicals NBP272781F
Description
MRPL15 Monoclonal specifically detects MRPL15 in Human, Mouse samples. It is validated for Immunohistochemistry.Specifications
MRPL15 | |
Monoclonal | |
FITC | |
PBS with 0.05% Sodium Azide | |
MRPL15 | |
Recombinant protein fragment corresponding to amino acids 28-296 of human MRPL15 (NP_054894) produced in E.coli. | |
0.1 mL | |
Primary | |
Human, Mouse | |
IgG2b |
Immunohistochemistry | |
OTI7A3 | |
Immunohistochemistry | |
L15mtHSPC145, mitochondrial ribosomal protein L15, MRP-L1539S ribosomal protein L15, mitochondrial, MRP-L7, RPML7 | |
Mouse | |
Affinity Purified | |
Core ESC Like Genes, Stem Cell Markers | |
29088.0 | |
Store at 4°C in the dark. |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction