Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ MRPL15 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA5109975
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA5109975 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA5109975 Supplier Invitrogen™ Supplier No. PA5109975
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Immunogen sequence: MAGPLQGGGA RALDLLRGLP RVSLANLKPN PGSKKPERRP RGRRRGRKCG RGHKGERQRG TRPRLGFEGG QTPFYIRIPK YGFNEGHSFR RQYKPLSLNR LQYLIDLGRV DPSQPIDLTQ LVNGRGVTIQ PLKRDYGVQL VEEGADTFTA KVNIEVQLAS ELAIAAIEKN GGVVTTAFYD PRSLDIVCKP VPFFLRGQPI PKRMLPPEEL VPYYTDAKNR GYLADPAKFP EARLELARKY GYILPDITKD ELFKMLCTRK DPRQIFFGLA PGWVVNMADK KILKPTDENL LKYYTS.

Mammalian mitochondrial ribosomal proteins are encoded by nuclear genes and help in protein synthesis within the mitochondrion. Mitochondrial ribosomes (mitoribosomes) consist of a small 28S subunit and a large 39S subunit. They have an estimated 75% protein to rRNA composition compared to prokaryotic ribosomes, where this ratio is reversed. Another difference between mammalian mitoribosomes and prokaryotic ribosomes is that the latter contain a 5S rRNA. Among different species, the proteins comprising the mitoribosome differ greatly in sequence, and sometimes in biochemical properties, which prevents easy recognition by sequence homology. This gene encodes a 39S subunit protein that belongs to the EcoL15 ribosomal protein family. A pseudogene corresponding to this gene is found on chromosome 15q.
TRUSTED_SUSTAINABILITY

Specifications

Antigen MRPL15
Applications ELISA, Western Blot
Classification Polyclonal
Concentration 1.95 mg/mL
Conjugate Unconjugated
Formulation PBS with 50% glycerol and 0.01% thimerosal; pH 7.3
Gene Mrpl15
Gene Accession No. Q9CPR5, Q9P015
Gene Alias 39S ribosomal protein L15, mitochondrial; HSPC145; L15mt; Mitochondrial large ribosomal subunit protein uL15m; mitochondrial ribosomal protein L15; Mrpl15; MRP-L15; MRP-L7; ribosomal protein, mitochondrial, L7; RPML7
Gene Symbols Mrpl15
Host Species Rabbit
Immunogen Recombinant fusion protein containing a sequence corresponding to amino acids 1-296 of human MRPL15 (NP_054894.1).
Purification Method Affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 27395, 29088
Target Species Human, Mouse
Content And Storage -20°C, Avoid Freeze/Thaw Cycles
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.