Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ MRPL16 Polyclonal Antibody

Rabbit Polyclonal Antibody

Supplier:  Invitrogen™ PA551009

Catalog No. PA551009


Only null left
Add to Cart

Description

Description

MRPL16 Polyclonal Antibody detects endogenous levels of MRPL16 protein.

Mammalian mitochondrial ribosomal proteins are encoded by nuclear genes and help in protein synthesis within the mitochondrion. Mitochondrial ribosomes (mitoribosomes) consist of a small 28S subunit and a large 39S subunit. They have an estimated 75% protein to rRNA composition compared to prokaryotic ribosomes, where this ratio is reversed. Another difference between mammalian mitoribosomes and prokaryotic ribosomes is that the latter contain a 5S rRNA. Among different species, the proteins comprising the mitoribosome differ greatly in sequence, and sometimes in biochemical properties, which prevents easy recognition by sequence homology. This gene encodes a 39S subunit protein. [provided by RefSeq, Jul 2008].
TRUSTED_SUSTAINABILITY
Specifications

Specifications

MRPL16
Polyclonal
Unconjugated
MRPL16
2310039D06Rik; 39S ribosomal protein L16, mitochondrial; L16mt; mitochondiral ribosomal protein L16; Mitochondrial large ribosomal subunit protein uL16m; mitochondrial ribosomal protein L16; Mrpl16; MRP-L16; PNAS-111; rm16
Rabbit
Antigen affinity chromatography
RUO
54948
-20°C, Avoid Freeze/Thaw Cycles
Liquid
ELISA, Immunohistochemistry, Western Blot
1 mg/mL
PBS with 0.5% BSA, 50% glycerol and 0.02% sodium azide; pH 7.4
Q9NX20
MRPL16
Synthesized peptide derived from the C-terminal region of human MRP-L16.
100 μL
Primary
Human
Antibody
IgG
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Product Certifications
Promotions

Promotions

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.