Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Invitrogen™ MRPL17 Polyclonal Antibody
Rabbit Polyclonal Antibody
Supplier: Invitrogen™ PA5103521
Description
Antibody detects endogenous levels of total MRPL17.
MRPL17 is a protein coding gene. Gene Ontology (GO) annotations related to this gene include ribosome; structural constituent of ribosome; translation.
Specifications
MRPL17 | |
Polyclonal | |
Unconjugated | |
MRPL17 | |
39S ribosomal protein L17, mitochondrial; L17mt; LIP2; LYST-interacting protein 2; LYST-interacting protein LIP2; Mitochondrial large ribosomal subunit protein bL17m; mitochondrial ribosomal protein L17; MRPL17; MRP-L17; MRP-L26; ribosomal protein, mitochondrial, L26; RPL17L; RPML26 | |
Rabbit | |
Affinity chromatography | |
RUO | |
27397, 63875 | |
-20°C | |
Liquid |
Western Blot, Immunocytochemistry | |
1 mg/mL | |
PBS with 50% glycerol and 0.02% sodium azide; pH 7.4 | |
Q9D8P4, Q9NRX2 | |
MRPL17 | |
A synthesized peptide derived from human MRPL17(Accession Q9NRX2), corresponding to amino acid residues A76-K126. | |
100 μL | |
Primary | |
Human, Mouse | |
Antibody | |
IgG |
Safety and Handling
WARNING: Cancer - www.P65Warnings.ca.gov
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction