Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ MRPL18 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA5109976
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA5109976 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA5109976 Supplier Invitrogen™ Supplier No. PA5109976
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Immunogen sequence: EAVAPEFTNR NPRNLELLGV ARKERGWATV WPNREFWHRL RVVKTQHHVE AFVEHLNGQV VVSASTREWA IKKHLYSTRN VVACESIGRV LAQRCLEAGI NFMVYQPTPW EASSDSIKRL QNAMTESGVM LREPRRIYE.

Mammalian mitochondrial ribosomal proteins are encoded by nuclear genes and help in protein synthesis within the mitochondrion. Mitochondrial ribosomes (mitoribosomes) consist of a small 28S subunit and a large 39S subunit. They have an estimated 75% protein to rRNA composition compared to prokaryotic ribosomes, where this ratio is reversed. Another difference between mammalian mitoribosomes and prokaryotic ribosomes is that the latter contain a 5S rRNA. Among different species, the proteins comprising the mitoribosome differ greatly in sequence, and sometimes in biochemical properties, which prevents easy recognition by sequence homology. This gene encodes a 39S subunit protein that belongs to the L18P ribosomal protein family. Three polymorphic sites exist in this gene, one of which is three nt in length which causes an extra aa near the N-terminus.
TRUSTED_SUSTAINABILITY

Specifications

Antigen MRPL18
Applications ELISA, Western Blot
Classification Polyclonal
Concentration 0.7 mg/mL
Conjugate Unconjugated
Formulation PBS with 50% glycerol and 0.01% thimerosal; pH 7.3
Gene Mrpl18
Gene Accession No. Q9CQL5, Q9H0U6
Gene Alias 1010001C05Rik; 39S ribosomal protein L18, mitochondrial; C79879; HSPC071; L18mt; Mitochondrial large ribosomal subunit protein uL18m; mitochondrial ribosomal protein L18; Mrpl18; MRP-L18
Gene Symbols Mrpl18
Host Species Rabbit
Immunogen Recombinant fusion protein containing a sequence corresponding to amino acids 42-180 of mouse Mrpl18 (NP_0805861).
Purification Method Affinity Chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 29074, 67681
Target Species Human, Mouse
Content And Storage -20°C, Avoid Freeze/Thaw Cycles
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.