Learn More
Invitrogen™ MRPL19 Polyclonal Antibody

Description
Antibody detects endogenous levels of total MRPL19.
Specifications
Specifications
| Antigen | MRPL19 |
| Applications | Immunohistochemistry (Paraffin), Western Blot, Immunocytochemistry |
| Classification | Polyclonal |
| Concentration | 1 mg/mL |
| Conjugate | Unconjugated |
| Formulation | PBS with 50% glycerol and 0.02% sodium azide |
| Gene | MRPL19 |
| Gene Accession No. | P49406, Q9D338 |
| Gene Alias | 39S ribosomal protein L15, mitochondrial; 39S ribosomal protein L19, mitochondrial; 9030416F12Rik; 9130412E02Rik; D6Ertd157e; KIAA0104; L15mt; L19mt; Mitochondrial large ribosomal subunit protein bL19m; mitochondrial ribosomal protein L19; MRPL15; MRP-L15; Mrpl19; MRP-L19; ribosomal protein, mitochondrial, L15; RLX1; RPML15 |
| Gene Symbols | MRPL19 |
| Show More |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.