Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Invitrogen™ MRPL19 Polyclonal Antibody
Rabbit Polyclonal Antibody
Supplier: Invitrogen™ PA565458
Description
Immunogen sequence: LRVTTADPYAS GKISQFLGIC IQRSGRGLGA TFILRNVIEG QGVEICFELY NPRVQEIQVV KLEKRLDDSL LYLRDALPEY ST Highest antigen sequence identity to the following orthologs - mouse 92%, rat 92%.
Mammalian mitochondrial ribosomal proteins are encoded by nuclear genes and help in protein synthesis within the mitochondrion. Mitochondrial ribosomes (mitoribosomes) consist of a small 28S subunit and a large 39S subunit. They have an estimated 75% protein to rRNA composition compared to prokaryotic ribosomes, where this ratio is reversed. Another difference between mammalian mitoribosomes and prokaryotic ribosomes is that the latter contain a 5S rRNA. Among different species, the proteins comprising the mitoribosome differ greatly in sequence, and sometimes in biochemical properties, which prevents easy recognition by sequence homology. This gene encodes a 39S subunit protein.
Specifications
MRPL19 | |
Polyclonal | |
Unconjugated | |
MRPL19 | |
39S ribosomal protein L15, mitochondrial; 39S ribosomal protein L19, mitochondrial; 9030416F12Rik; 9130412E02Rik; D6Ertd157e; KIAA0104; L15mt; L19mt; Mitochondrial large ribosomal subunit protein bL19m; mitochondrial ribosomal protein L19; MRPL15; MRP-L15; Mrpl19; MRP-L19; ribosomal protein, mitochondrial, L15; RLX1; RPML15 | |
Rabbit | |
Antigen affinity chromatography | |
RUO | |
9801 | |
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
Liquid |
Immunocytochemistry | |
0.1 mg/mL | |
PBS with 40% glycerol and 0.02% sodium azide; pH 7.2 | |
P49406 | |
MRPL19 | |
Recombinant Human MRPL19. Recombinant protein control fragment (Product #RP-106136). | |
100 μL | |
Primary | |
Human | |
Antibody | |
IgG |
Safety and Handling
WARNING: Cancer - www.P65Warnings.ca.gov
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction