Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
MRPL20 Antibody, mFluor Violet 450 SE, Novus Biologicals™

Rabbit Polyclonal Antibody
Supplier: Novus Biologicals NBP298645MFV450
Description
MRPL20 Polyclonal antibody specifically detects MRPL20 in Human samples. It is validated for Western Blot, Immunoprecipitation, Immunohistochemistry (Paraffin)Specifications
MRPL20 | |
Polyclonal | |
Western Blot, Immunoprecipitation, Immunohistochemistry-Paraffin | |
39S ribosomal protein L20, mitochondrial, FLJ10024, L20mt, MGC4779, MGC74465, mitochondrial ribosomal protein L20, MRP-L20 | |
Produced in rabbits immunized with E. coli-derived Human MRPL20 fragment. | |
0.1 mL | |
Primary | |
Human | |
Purified |
Western Blot, Immunoprecipitation, Immunohistochemistry (Paraffin) | |
mFluor Violet 450 SE | |
50mM Sodium Borate | |
Rabbit | |
Antigen and protein A Affinity-purified | |
RUO | |
55052 | |
Store at 4C in the dark. | |
IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction