Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ MRPL20 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA561417
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA561417 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA561417 Supplier Invitrogen™ Supplier No. PA561417
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Immunogen sequence: ASQEHGLKYP ALIGNLVKCQ VELNRKVLAD LAIYEPKTFK SLAALASRRR HEGFAAALGD GKEPEGIFSR VVQYH Highest antigen sequence identity to the following orthologs: Mouse - 91%, Rat - 91%.

Mammalian mitochondrial ribosomal proteins are encoded by nuclear genes and help in protein synthesis within the mitochondrion. Mitochondrial ribosomes (mitoribosomes) consist of a small 28S subunit and a large 39S subunit. They have an estimated 75% protein to rRNA composition compared to prokaryotic ribosomes, where this ratio is reversed. Another difference between mammalian mitoribosomes and prokaryotic ribosomes is that the latter contain a 5S rRNA. Among different species, the proteins comprising the mitoribosome differ greatly in sequence, and sometimes in biochemical properties, which prevents easy recognition by sequence homology. This gene encodes a 39S subunit protein. A pseudogene corresponding to this gene is found on chromosome 21q. [provided by RefSeq, Jul 2008].
TRUSTED_SUSTAINABILITY

Specifications

Antigen MRPL20
Applications Immunohistochemistry (Paraffin), Western Blot, Immunocytochemistry
Classification Polyclonal
Concentration 0.2 mg/mL
Conjugate Unconjugated
Formulation PBS with 40% glycerol and 0.02% sodium azide; pH 7.2
Gene MRPL20
Gene Accession No. Q9BYC9
Gene Alias 2610008D01Rik; 39S ribosomal protein L20, mitochondrial; 4930425I20Rik; L20mt; Mitochondrial large ribosomal subunit protein bL20m; mitochondrial ribosomal protein L20; MRPL20; MRP-L20
Gene Symbols MRPL20
Host Species Rabbit
Immunogen Recombinant protein corresponding to Human MRPL20. Recombinant protein control fragment (Product #RP-100655).
Purification Method Antigen affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 55052
Target Species Human
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.