Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
MRPL23 Polyclonal Antibody
MRPL23 Polyclonal Antibody
Supplier: Thermo Scientific PA578153
Description
MRPL23 Polyclonal Antibody for IHC (P)

Specifications
MRPL23 | |
Polyclonal | |
Unconjugated | |
MRPL23 | |
39S ribosomal protein L23, mitochondrial; FLJ45387; L23 mitochondrial-related protein; L23mrp; L23mt; Mitochondrial large ribosomal subunit protein uL23m; mitochondrial ribosomal protein L23; Mrpl23; MRP-L23; Ribosomal protein L23-like; ribosomal protein L23-related product homolog; ribosomal protein related to L23 (mitochondrial); rL23MRP; Rpl23; RPL23L; Rpl23-rs | |
Rabbit | |
Antigen Affinity Chromatography | |
RUO | |
6150, 64360 | |
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
Liquid |
Immunohistochemistry (Paraffin) | |
1 mg/mL | |
0.1M tris glycine with 10% glycerol, 0.1M glycine and 0.01% thimerosal; pH 7 | |
Q16540, Q63750 | |
MRPL23 | |
Recombinant protein encompassing a sequence within the center region of human MRPL23. The exact sequence is proprietary. | |
100 μL | |
Primary | |
Human, Rat | |
Antibody | |
IgG |
Safety and Handling
WARNING: Cancer - www.P65Warnings.ca.gov
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction