Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ MRPL24 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA5101359
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA5101359 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA5101359 Supplier Invitrogen™ Supplier No. PA5101359
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Antibody detects endogenous levels of total MRPL24.

Mammalian mitochondrial ribosomal proteins are encoded by nuclear genes and help in protein synthesis within the mitochondrion. Mitochondrial ribosomes consist of a small 28S subunit and a large 39S subunit. They have an estimated 75% protein to rRNA composition compared to prokaryotic ribosomes, where this ratio is reversed. Another difference between mammalian mitoribosomes and prokaryotic ribosomes is that the latter contain a 5S rRNA. Among different species, the proteins comprising the mitoribosome differ greatly in sequence, and sometimes in biochemical properties, which prevents easy recognition by sequence homology. This gene encodes a 39S subunit protein which is more than twice the size of its E. coli counterpart. Sequence analysis identified two transcript variants that encode the same protein.
TRUSTED_SUSTAINABILITY

Specifications

Antigen MRPL24
Applications Immunohistochemistry (Paraffin), Western Blot, Immunocytochemistry
Classification Polyclonal
Concentration 1 mg/mL
Conjugate Unconjugated
Formulation PBS with 50% glycerol and 0.02% sodium azide; pH 7.4
Gene mrpl24
Gene Accession No. Q66H47, Q96A35, Q9CQ06
Gene Alias 2010005E08Rik; 2810470K06Rik; 39S ribosomal protein L24, mitochondrial; 6720473G22Rik; AA407670; FLJ20917; hypothetical protein LOC532203; L24mt; MGC22737; MGC9831; Mitochondrial large ribosomal subunit protein uL24m; mitochondrial ribosomal protein L24; MRP-L18; MRPL24; MRP-L24; MRPL24 protein
Gene Symbols mrpl24
Host Species Rabbit
Immunogen A synthesized peptide derived from human MRPL24(Accession Q96A35), corresponding to amino acid residues D103-I153.
Purification Method Affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 295224, 67707, 79590
Target Species Human, Mouse, Rat
Content And Storage -20°C
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.