Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ MRPL27 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA554402
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA554402 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA554402 Supplier Invitrogen™ Supplier No. PA554402
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Immunogen sequence: HPGAHVGVGK NKCLYALEEG IVRYTKEVYV PHPRNTEAVD LITRLPKGAV LYKTFVHVVP AKPEGTFKLV AML Highest antigen sequence identity to the following orthologs: Mouse - 89%, Rat - 89%.

Mammalian mitochondrial ribosomal proteins are encoded by nuclear genes and help in protein synthesis within the mitochondrion. Mitochondrial ribosomes (mitoribosomes) consist of a small 28S subunit and a large 39S subunit. They have an estimated 75% protein to rRNA composition compared to prokaryotic ribosomes, where this ratio is reversed. Another difference between mammalian mitoribosomes and prokaryotic ribosomes is that the latter contain a 5S rRNA. Among different species, the proteins comprising the mitoribosome differ greatly in sequence, and sometimes in biochemical properties, which prevents easy recognition by sequence homology. MRPL27 encodes a 39S subunit protein.
TRUSTED_SUSTAINABILITY

Specifications

Antigen MRPL27
Applications Immunohistochemistry (Paraffin)
Classification Polyclonal
Concentration 0.1 mg/mL
Conjugate Unconjugated
Formulation PBS with 40% glycerol and 0.02% sodium azide; pH 7.2
Gene MRPL27
Gene Accession No. Q9P0M9
Gene Alias 39S ribosomal protein L27, mitochondrial; A630055F16Rik; D11Moh47; D18Ertd643e; HSPC250; L27mt; Mitochondrial large ribosomal subunit protein bL27m; mitochondrial ribosomal protein L27; Mrpl27; MRP-L27
Gene Symbols MRPL27
Host Species Rabbit
Immunogen Recombinant protein corresponding to Human MRPL27. Recombinant protein control fragment (Product #RP-93208).
Purification Method Antigen affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 51264
Target Species Human
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.