Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ MRPL28 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA5103523
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA5103523 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA5103523 Supplier Invitrogen™ Supplier No. PA5103523
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Antibody detects endogenous levels of total MRPL28.

MRP-L28 (39S ribosomal protein L28, melanoma-associated antigen recognized by T lymphocytes) is a mitochondrial protein that belongs to the ribosomal protein L28P family. MRP-L28 potentially represents an important therapeutic reagent for HLA-A24 (A24) patients as this antigen is recognized by tumor-infiltrating lymphocyte (TIL) 1290, which targets the A24 serotype. HLAA24 (A24) is an HLA-A serotype that identifies the more common HLA-A24 gene products. A24 is a split antigen that is also recognized by the A9 broad antigen type and the similar A23 types. A24 is common in Austronesia and has one of the highest A allele frequencies for a number of peoples, including Papua New Guineans, indigeonous Taiwanese (eastern tribals), Yupik and Greenland Eskimos. MRP-L28 is found in a variety of normal tissues including spleen, testis, thymus, liver, kidney, brain, adrenal, lung and retinal tissue.
TRUSTED_SUSTAINABILITY

Specifications

Antigen MRPL28
Applications Immunohistochemistry (Paraffin), Western Blot
Classification Polyclonal
Concentration 1 mg/mL
Conjugate Unconjugated
Formulation PBS with 50% glycerol and 0.02% sodium azide; pH 7.4
Gene MRPL28
Gene Accession No. Q13084, Q9D1B9
Gene Alias 1110015G04Rik; 39S ribosomal protein L28, mitochondrial; HGNC6756; L28mt; MAAT 1; MAAT1; Melanoma antigen p15; melanoma-associated antigen recognised by cytotoxic T lymphocytes; melanoma-associated antigen recognized by T lymphocytes; melanoma-associated antigen recognized by T-lymphocytes; MGC8499; Mitochondrial large ribosomal subunit protein bL28m; mitochondrial ribosomal protein L28; MRP L28; MRPL 28; MRPL28; MRP-L28; p15
Gene Symbols MRPL28
Host Species Rabbit
Immunogen A synthesized peptide derived from human MRPL28(Accession Q13084), corresponding to amino acid residues L146-I196.
Purification Method Affinity Chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 10573, 68611
Target Species Human, Mouse, Monkey
Content And Storage -20°C
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.