Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
MRPL3 Antibody, mFluor Violet 500 SE, Novus Biologicals™

Rabbit Polyclonal Antibody
Supplier: Novus Biologicals NBP297939MFV500
Description
MRPL3 Polyclonal antibody specifically detects MRPL3 in Human samples. It is validated for Western BlotSpecifications
MRPL3 | |
Polyclonal | |
Western Blot | |
39S ribosomal protein L3, mitochondrial, mitochondrial ribosomal protein L3, MRL3mitochondrial, L3, RPML3 | |
Produced in rabbits immunized with E. coli-derived Human MRPL3 fragment. | |
0.1 mL | |
Primary | |
Human | |
Purified |
Western Blot | |
mFluor Violet 500 SE | |
50mM Sodium Borate | |
Rabbit | |
Antigen and protein A Affinity-purified | |
RUO | |
11222 | |
Store at 4C in the dark. | |
IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction