Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Invitrogen™ MRPL3 Polyclonal Antibody
Rabbit Polyclonal Antibody
Supplier: Invitrogen™ PA521892
Description
Recommended positive controls: 293T, A431. Predicted reactivity: Chimpanzee (100%). Store product as a concentrated solution. Centrifuge briefly prior to opening the vial.
Mammalian mitochondrial ribosomal proteins are encoded by nuclear genes and help in protein synthesis within the mitochondrion. Mitochondrial ribosomes (mitoribosomes) consist of a small 28S subunit and a large 39S subunit. They have an estimated 75% protein to rRNA composition compared to prokaryotic ribosomes, where this ratio is reversed. Another difference between mammalian mitoribosomes and prokaryotic ribosomes is that the latter contain a 5S rRNA. Among different species, the proteins comprising the mitoribosome differ greatly in sequence, and sometimes in biochemical properties, which prevents easy recognition by sequence homology. This gene encodes a 39S subunit protein that belongs to the L3P ribosomal protein family. A pseudogene corresponding to this gene is found on chromosome 13q.
Specifications
MRPL3 | |
Polyclonal | |
Unconjugated | |
MRPL3 | |
2010320L16Rik; 39S ribosomal protein L3, mitochondrial; 5930422H18Rik; AA407889; COXPD9; L3mt; mitochondrial 60S ribosomal protein L3; Mitochondrial large ribosomal subunit protein uL3m; mitochondrial ribosomal protein L3; MRL3; Mrpl3; MRP-L3; RPML3 | |
Rabbit | |
Antigen Affinity Chromatography | |
RUO | |
11222 | |
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
Liquid |
Western Blot | |
1 mg/mL | |
0.1M tris glycine with 10% glycerol and 0.01% thimerosal; pH 7 | |
P09001 | |
MRPL3 | |
Synthetic peptide corresponding to a region within amino acids 286 and 348 of MRPL3 (Uniprot ID#P09001). | |
100 μL | |
Primary | |
Human | |
Antibody | |
IgG |
Safety and Handling
WARNING: Cancer - www.P65Warnings.ca.gov
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction