Learn More
Invitrogen™ MRPL32 Polyclonal Antibody

Description
Immunogen sequence: VLRNYWERLL RKLPQSRPGF PSPPWGPALA VQGPAMFTEP ANDTSGSKEN SSLLDSIFWM AAPKNRRTIE VNRCRRRNPQ KLIKVKNNID VCPECGHLKQ KHVLCAYCYE KVCKETAEIR RQIGKQEGGP FKAPTIETVV LYTGETPSEQ DQGKRIIERD RKRPSWFTQN.
Specifications
Specifications
| Antigen | MRPL32 |
| Applications | ELISA, Western Blot, Immunocytochemistry |
| Classification | Polyclonal |
| Concentration | 0.78 mg/mL |
| Conjugate | Unconjugated |
| Formulation | PBS with 50% glycerol and 0.02% sodium azide; pH 7.3 |
| Gene | MRPL32 |
| Gene Accession No. | Q9BYC8, Q9DCI9 |
| Gene Alias | 0610033O15Rik; 39S ribosomal protein L32, mitochondrial; bMRP-59b; Heart-expressed gene 1 protein; Heg1; HSPC283; L32mt; Mitochondrial large ribosomal subunit protein bL32m; mitochondrial ribosomal protein L32; MRPL32; MRP-L32 |
| Gene Symbols | MRPL32 |
| Show More |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.