Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
MRPL34 Antibody, DyLight 594, Novus Biologicals™

Rabbit Polyclonal Antibody
Supplier: Novus Biologicals NBP297371DL594
Description
MRPL34 Polyclonal specifically detects MRPL34 in Human samples. It is validated for Immunocytochemistry/Immunofluorescence.Specifications
MRPL34 | |
Polyclonal | |
Immunocytochemistry/Immunofluorescence | |
L34mtMGC24974, MGC2633, mitochondrial ribosomal protein L34, MRP-L34,39S ribosomal protein L34, mitochondrial | |
Produced in rabbits immunized with E. coli-derived Human MRPL34 fragment. | |
0.1 mL | |
Stem Cell Markers | |
64981 | |
Store at 4°C in the dark. | |
IgG |
Immunofluorescence | |
DyLight 594 | |
50mM Sodium Borate | |
Rabbit | |
Antigen Affinity-purified | |
RUO | |
Primary | |
Human | |
Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction