Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Invitrogen™ MRPL34 Polyclonal Antibody
Rabbit Polyclonal Antibody
Supplier: Invitrogen™ PA568064
Description
Predicted to react with Mouse samples.
Mammalian mitochondrial ribosomal proteins are encoded by nuclear genes and help in protein synthesis within the mitochondrion. Mitochondrial ribosomes (mitoribosomes) consist of a small 28S subunit and a large 39S subunit. They have an estimated 75% protein to rRNA composition compared to prokaryotic ribosomes, where this ratio is reversed. Another difference between mammalian mitoribosomes and prokaryotic ribosomes is that the latter contain a 5S rRNA. Among different species, the proteins comprising the mitoribosome differ greatly in sequence, and sometimes in biochemical properties, which prevents easy recognition by sequence homology. This gene encodes a 39S subunit protein.
Specifications
MRPL34 | |
Polyclonal | |
Unconjugated | |
Mrpl34 | |
0610007O17Rik; 39S ribosomal protein L34, mitochondrial; D8Bwg1484e; L34mt; Mitochondrial large ribosomal subunit protein bL34m; mitochondrial ribosomal protein L34; MRPL34; MRP-L34; RM34 | |
Rabbit | |
Affinity chromatography | |
RUO | |
64981, 94065 | |
-20°C | |
Liquid |
Immunohistochemistry, Immunohistochemistry (Paraffin), Western Blot | |
1 mg/mL | |
PBS with 30% glycerol and 0.02% sodium azide; pH 7.3 | |
Q99N91, Q9BQ48 | |
Mrpl34 | |
Synthesized peptide derived from human MRPL34. | |
100 μL | |
Primary | |
Human, Mouse | |
Antibody | |
IgG |
Safety and Handling
WARNING: Cancer - www.P65Warnings.ca.gov
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction