Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

MRPL36 Rabbit anti-Human, Polyclonal, Proteintech
SDP

Catalog No. 501731066 Shop All Proteintech Group Inc Products
Encompass_Preferred
Change view
Click to view available options
Quantity:
150 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
50-173-1066 150 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 50-173-1066 Supplier Proteintech Group Inc Supplier No. 166251AP150UL
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Mammalian mitochondrial ribosomal proteins are encoded by nuclear genes and help in protein synthesis within the mitochondrion. Mitochondrial ribosomes (mitoribosomes) consist of a small 28S subunit and a large 39S subunit. They have an estimated 75% protein to rRNA composition compared to prokaryotic ribosomes, where this ratio is reversed. Another difference between mammalian mitoribosomes and prokaryotic ribosomes is that the latter contain a 5S rRNA. Among different species, the proteins comprising the mitoribosome differ greatly in sequence, and sometimes in biochemical properties, which prevents easy recognition by sequence homology. MRPL36 encodes a 39S subunit protein. A pseudogene corresponding to this gene is found on chromosome 2p.

Specifications

Antigen MRPL36
Applications Immunohistochemistry (Paraffin)
Classification Polyclonal
Concentration 0.24 mg/mL
Conjugate Unconjugated
Formulation PBS with 50% glycerol and 0.02% sodium azide; pH 7.3
Gene MRPL36
Gene Accession No. Q9P0J6
Gene Alias BRCA1 interacting protein 1, BRIP1, L36mt, MRP L36, MRPL36, PRPL36, RPMJ
Gene Symbols MRPL36
Host Species Rabbit
Immunogen MRPL36 Fusion Protein Ag9925
Purification Method Antigen Affinity Chromatography
Quantity 150 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 64979
Target Species Human
Content And Storage -20°C
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.