Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ MRPL37 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA544650
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA544650 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA544650 Supplier Invitrogen™ Supplier No. PA544650
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Peptide sequence: VRSTRKSEPP PLDRVYEIPG LEPITFAGKM HFVPWLARPI FPPWDRGYKD Sequence homology: Cow: 100%; Dog: 86%; Guinea Pig: 100%; Horse: 100%; Human: 100%; Mouse: 100%; Rabbit: 100%; Rat: 100%.

Mammalian mitochondrial ribosomal proteins are encoded by nuclear genes and help in protein synthesis within the mitochondrion. Mitochondrial ribosomes (mitoribosomes) consist of a small 28S subunit and a large 39S subunit.
TRUSTED_SUSTAINABILITY

Specifications

Antigen MRPL37
Applications Western Blot
Classification Polyclonal
Concentration 0.5 mg/mL
Conjugate Unconjugated
Formulation PBS with 2% sucrose and 0.09% sodium azide
Gene MRPL37
Gene Accession No. Q9BZE1
Gene Alias 2300004O14Rik; 39S ribosomal protein L2, mitochondrial; 39S ribosomal protein L37, mitochondrial; AI132596; HSPC235; L2mt; L37mt; MGC878; Mitochondrial large ribosomal subunit protein mL37; mitochondrial ribosomal protein L37; MRPL2; MRP-L2; MRPL37; MRP-L37; ribosomal protein, mitochondrial, L2; RPML2
Gene Symbols MRPL37
Host Species Rabbit
Immunogen Synthetic peptide directed towards the N-terminal of human MRPL37 (aa 35-84).
Purification Method Affinity Chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 51253
Target Species Human
Content And Storage -20°C, Avoid Freeze/Thaw Cycles
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.