Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ MRPL37 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA555274
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA555274 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA555274 Supplier Invitrogen™ Supplier No. PA555274
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Immunogen sequence: LTKTKLIEGL PEKVLSLVDD PRNHIENQDE CVLNVISHAR LWQTTEEIPK RETYCPVIVD NLIQLCKSQI LKHP Highest antigen sequence identity to the following orthologs: Mouse - 84%, Rat - 81%.

Mammalian mitochondrial ribosomal proteins are encoded by nuclear genes and help in protein synthesis within the mitochondrion. Mitochondrial ribosomes (mitoribosomes) consist of a small 28S subunit and a large 39S subunit.
TRUSTED_SUSTAINABILITY

Specifications

Antigen MRPL37
Applications Immunohistochemistry (Paraffin), Immunocytochemistry
Classification Polyclonal
Concentration 0.1 mg/mL
Conjugate Unconjugated
Formulation PBS with 40% glycerol and 0.02% sodium azide; pH 7.2
Gene MRPL37
Gene Accession No. Q9BZE1
Gene Alias 2300004O14Rik; 39S ribosomal protein L2, mitochondrial; 39S ribosomal protein L37, mitochondrial; AI132596; HSPC235; L2mt; L37mt; MGC878; Mitochondrial large ribosomal subunit protein mL37; mitochondrial ribosomal protein L37; MRPL2; MRP-L2; MRPL37; MRP-L37; ribosomal protein, mitochondrial, L2; RPML2
Gene Symbols MRPL37
Host Species Rabbit
Immunogen Recombinant protein corresponding to Human MRPL37. Recombinant protein control fragment (Product #RP-94068).
Purification Method Antigen Affinity Chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 51253
Target Species Human
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.