Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ MRPL38 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA5109981
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA5109981 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA5109981 Supplier Invitrogen™ Supplier No. PA5109981
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Immunogen sequence: MPNSDIDLSN LERLEKYRSF DRYRRRAEQE AQAPHWWRTY REYFGEKTDP KEKIDIGLPP PKVSRTQQLL ERKQAIQELR ANVEEERAAR LRTASVPLDA VRAEWERTCG PYHKQRLAEY YGLYRDLFHG ATFVPRVPLH VAYAVGEDDL MPVYCGNEVT PTEAAQAPEV TYEAEEGSLW TLLLTSLDGH LLEPDAEYLH WLLTNIPGNR VAEGQVTCPY LPPFPARGSG IHRLAFLLFK QDQPIDFSED ARPSPCYQLA QRTFRTFDFY KKHQETMTPA GLSFFQCRWD DSVTYIFHQL LDMREPVFEF VRPPPYHPKQ KRFPHRQPLR YLDRYRDSHE PTYGIY.

Mammalian mitochondrial ribosomal proteins are encoded by nuclear genes and help in protein synthesis within the mitochondrion. Mitochondrial ribosomes (mitoribosomes) consist of a small 28S subunit and a large 39S subunit. They have an estimated 75% protein to rRNA composition compared to prokaryotic ribosomes, where this ratio is reversed. Another difference between mammalian mitoribosomes and prokaryotic ribosomes is that the latter contain a 5S rRNA. Among different species, the proteins comprising the mitoribosome differ greatly in sequence, and sometimes in biochemical properties, which prevents easy recognition by sequence homology. This gene encodes a 39S subunit protein.
TRUSTED_SUSTAINABILITY

Specifications

Antigen MRPL38
Applications ELISA, Immunohistochemistry (Paraffin), Western Blot
Classification Polyclonal
Concentration 0.56 mg/mL
Conjugate Unconjugated
Formulation PBS with 50% glycerol and 0.01% thimerosal; pH 7.3
Gene MRPL38
Gene Accession No. Q5PQN9, Q8K2M0, Q96DV4
Gene Alias 1110036N21Rik; 39S ribosomal protein L38, mitochondrial; 4733401F03Rik; HSPC262; L38mt; Mitochondrial large ribosomal subunit protein mL38; mitochondrial ribosomal protein L38; MRP-L3; MRPL38; MRP-L38; R75240; ribosomal protein, mitochondrial, L3; rm38; Rpml3
Gene Symbols MRPL38
Host Species Rabbit
Immunogen Recombinant fusion protein containing a sequence corresponding to amino acids 35-380 of human MRPL38 (NP_1158672).
Purification Method Affinity Chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 303685, 60441, 64978
Target Species Human, Mouse, Rat
Content And Storage -20°C, Avoid Freeze/Thaw Cycles
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.