Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
MRPL39 Polyclonal Antibody
MRPL39 Polyclonal Antibody
Supplier: Thermo Scientific PA532108
Description
MRPL39 Polyclonal Antibody for Western Blot

Specifications
MRPL39 | |
Polyclonal | |
Unconjugated | |
MRPL39 | |
39S ribosomal protein L39, mitochondrial; 39S ribosomal protein L5, mitochondrial; C21orf8; C21orf92; L39mt; L5mt; Mitochondrial large ribosomal subunit protein mL39; mitochondrial ribosomal protein L39; MRPL39; MRP-L39; MRPL5; MRP-L5; MSTP003; ORF22; PRED22; PRED66; ribosomal protein, mitochondrial, L5; RPML5 | |
Rabbit | |
Antigen affinity chromatography | |
RUO | |
54148 | |
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
Liquid |
Western Blot | |
0.77 mg/mL | |
0.1M tris glycine with 20% glycerol and 0.01% thimerosal; pH 7 | |
Q9NYK5 | |
MRPL39 | |
Recombinant protein fragment corresponding to a region within amino acids 115 and 288 of Human MRPL39. | |
100 μL | |
Primary | |
Human | |
Antibody | |
IgG |
Safety and Handling
WARNING: Cancer - www.P65Warnings.ca.gov
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction