Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
MRPL40 Antibody, Novus Biologicals™

Rabbit Polyclonal Antibody has been used in 3 publications
$254.22 - $665.16
Specifications
| Antigen | MRPL40 |
|---|---|
| Applications | Western Blot, Immunohistochemistry, Immunocytochemistry, Immunofluorescence, Immunohistochemistry (Paraffin), KnockDown |
| Classification | Polyclonal |
| Conjugate | Unconjugated |
| Host Species | Rabbit |
Description
MRPL40 Polyclonal specifically detects MRPL40 in Human, Mouse, Rat samples. It is validated for Western Blot, Immunohistochemistry, Immunocytochemistry/Immunofluorescence, Immunohistochemistry-Paraffin, Knockdown Validated.Specifications
| MRPL40 | |
| Polyclonal | |
| Rabbit | |
| Human, Mouse, Rat | |
| FLJ41774, L40mt, MGC9400, mitochondrial ribosomal protein L40, MRP-L22,39S ribosomal protein L40, mitochondrial, MRP-L40, NLVCFURIM, nuclear localization signal containing protein deleted in velocardiofacialsyndrome, nuclear localization signal deleted in velocardiofacial syndrome, Nuclear localization signal-containing protein deleted in velocardiofacialsyndrome, Up-regulated in metastasis | |
| MRPL40 | |
| IgG | |
| Affinity Purified | |
| Specificity of antibody verified on a Protein Array containing target protein plus 383 other non-specific proteins. |
| Western Blot, Immunohistochemistry, Immunocytochemistry, Immunofluorescence, Immunohistochemistry (Paraffin), KnockDown | |
| Unconjugated | |
| RUO | |
| PBS (pH 7.2) and 40% Glycerol with 0.02% Sodium Azide | |
| 64976 | |
| This antibody was developed against Recombinant Protein corresponding to amino acids:TQLRETQELIPIEDFITPLKFLDKARERPQVELTFEETERRALLLKKWSLYKQQERKMERDTIRAMLEAQQEALEELQLESPKLHAEAIKRDPNLFPFEKEGPHYTPPIPNYQPPEGRYNDITK | |
| Primary | |
| Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles. |
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title