Learn More
Invitrogen™ MRPL40 Polyclonal Antibody

Description
MRPL40 Polyclonal Antibody detects endogenous levels of MRPL40 protein.
Specifications
Specifications
| Antigen | MRPL40 |
| Applications | Immunohistochemistry, Western Blot |
| Classification | Polyclonal |
| Concentration | 1 mg/mL |
| Conjugate | Unconjugated |
| Formulation | PBS with 0.5% BSA, 50% glycerol and 0.02% sodium azide; pH 7.4 |
| Gene | MRPL40 |
| Gene Accession No. | Q9NQ50 |
| Gene Alias | 39S ribosomal protein L40, mitochondrial; L40mt; Mitochondrial large ribosomal subunit protein mL40; mitochondrial ribosomal protein L40; MRPL22; MRP-L22; Mrpl40; MRP-L40; Nlvcf; nuclear localization signal protein absent in velo-cardio-facial patients; nuclear localization signal-containing protein deleted in velocardiofacial syndrome; Nuclear localization signal-containing protein deleted in velocardiofacial syndrome homolog; up-regulated in metastasis; URIM |
| Gene Symbols | MRPL40 |
| Show More |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.