Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Invitrogen™ MRPL40 Polyclonal Antibody

Rabbit Polyclonal Antibody
Supplier: Invitrogen™ PA5109982
Description
Immunogen sequence: MRSEPLRKKK KVDPKKDQEA KERLKRKIRK LEKATQELIP IEDFITPLKF LDKARERPQV ELTFEETERR ALLLKKWSLY KQQERKMERD TIRAMLEAQQ EALEELQLES PKLHAEAIKR DPNLFPFEKE GPHYTPPIPN YQPPEGRYND ITKVYTQVEF KR.
Mammalian mitochondrial ribosomal proteins are encoded by nuclear genes and help in protein synthesis within the mitochondrion. Mitochondrial ribosomes (mitoribosomes) consist of a small 28S subunit and a large 39S subunit. They have an estimated 75% protein to rRNA composition compared to prokaryotic ribosomes, where this ratio is reversed. Another difference between mammalian mitoribosomes and prokaryotic ribosomes is that the latter contain a 5S rRNA. Among different species, the proteins comprising the mitoribosome differ greatly in sequence, and sometimes in biochemical properties, which prevents easy recognition by sequence homology. This gene encodes a 39S subunit protein. Deletions in this gene may contribute to the etiology of velo-cardio-facial syndrome and DiGeorge syndrome.
Specifications
| MRPL40 | |
| Polyclonal | |
| Unconjugated | |
| MRPL40 | |
| 39S ribosomal protein L40, mitochondrial; L40mt; Mitochondrial large ribosomal subunit protein mL40; mitochondrial ribosomal protein L40; MRPL22; MRP-L22; Mrpl40; MRP-L40; Nlvcf; nuclear localization signal protein absent in velo-cardio-facial patients; nuclear localization signal-containing protein deleted in velocardiofacial syndrome; Nuclear localization signal-containing protein deleted in velocardiofacial syndrome homolog; up-regulated in metastasis; URIM | |
| Rabbit | |
| Affinity Chromatography | |
| RUO | |
| 64976 | |
| -20°C, Avoid Freeze/Thaw Cycles | |
| Liquid |
| ELISA, Western Blot | |
| 0.7 mg/mL | |
| PBS with 50% glycerol and 0.01% thimerosal; pH 7.3 | |
| Q9NQ50 | |
| MRPL40 | |
| Recombinant fusion protein containing a sequence corresponding to amino acids 45-206 of human MRPL40 (NP_0037672). | |
| 100 μL | |
| Primary | |
| Human | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction