Learn More
Invitrogen™ MRPL42 Polyclonal Antibody

Description
Immunogen sequence: YEHTKPIPRP DPVHNNEETH DQVLKTRLEE KVEHLEEGPM IEQLSKMFFT TKHRWYPHGR YHRCRKNLNP PKD Highest antigen sequence identity to the following orthologs: Mouse - 73%, Rat - 77%.
Specifications
Specifications
| Antigen | MRPL42 |
| Applications | Immunohistochemistry (Paraffin), Immunocytochemistry |
| Classification | Polyclonal |
| Concentration | 1.3 mg/mL |
| Conjugate | Unconjugated |
| Formulation | PBS with 40% glycerol and 0.02% sodium azide; pH 7.2 |
| Gene | MRPL42 |
| Gene Accession No. | Q9Y6G3 |
| Gene Alias | 2700009F22Rik; 28S ribosomal protein L42, mitochondrial; 28S ribosomal protein S32, mitochondrial; 2900055D03Rik; 39S ribosomal protein L31, mitochondrial; 39S ribosomal protein L42, mitochondrial; D10Ertd322e; HSPC204; L31mt; L42MT; mitochndrial ribosomal protein L42; Mitochondrial 28S ribosomal protein S32, mitochondrial; Mitochondrial large ribosomal subunit protein mL42; mitochondrial ribosomal protein L42; MRPL31; MRP-L31; MRPL42; MRP-L42; MRPS32; MRP-S32; PTD007; RPML31; S32mt |
| Gene Symbols | MRPL42 |
| Show More |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.