Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
MRPL42 Polyclonal Antibody
MRPL42 Polyclonal Antibody
Supplier: Thermo Scientific PA575888
Description
MRPL42 Polyclonal Antibody for Western Blot

Specifications
MRPL42 | |
Polyclonal | |
Unconjugated | |
MRPL42 | |
2700009F22Rik; 28S ribosomal protein L42, mitochondrial; 28S ribosomal protein S32, mitochondrial; 2900055D03Rik; 39S ribosomal protein L31, mitochondrial; 39S ribosomal protein L42, mitochondrial; D10Ertd322e; HSPC204; L31mt; L42MT; mitochndrial ribosomal protein L42; Mitochondrial 28S ribosomal protein S32, mitochondrial; Mitochondrial large ribosomal subunit protein mL42; mitochondrial ribosomal protein L42; MRPL31; MRP-L31; MRPL42; MRP-L42; MRPS32; MRP-S32; PTD007; RPML31; S32mt | |
Rabbit | |
Antigen Affinity Chromatography | |
RUO | |
28977, 299743, 67270 | |
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
Liquid |
Western Blot | |
1 mg/mL | |
PBS with 50% glycerol and 0.02% sodium azide; pH 7.2 | |
P0C2B9, Q9CPV3, Q9Y6G3 | |
MRPL42 | |
Synthetic peptide corresponding to residues in Human MRPL42. | |
100 μL | |
Primary | |
Human, Mouse, Rat | |
Antibody | |
IgG |
Safety and Handling
WARNING: Cancer - www.P65Warnings.ca.gov
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction