Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ MRPL43 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA5116773
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA5116773 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA5116773 Supplier Invitrogen™ Supplier No. PA5116773
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Positive Samples: A-549, PC-3, 293T Immunogen sequence: LNGAVREESI HCKSVEEIST LVQKLADQSG LDVIRIRKPF HTDNPSIQGQ WHPFTNKPTT FRGLRPREVQ DPAPAQ.

Mammalian mitochondrial ribosomal proteins are encoded by nuclear genes and help in protein synthesis within the mitochondrion. Mitochondrial ribosomes (mitoribosomes) consist of a small 28S subunit and a large 39S subunit. They have an estimated 75% protein to rRNA composition compared to prokaryotic ribosomes, where this ratio is reversed. Another difference between mammalian mitoribosomes and prokaryotic ribosomes is that the latter contain a 5S rRNA. Among different species, the proteins comprising the mitoribosome differ greatly in sequence, and sometimes in biochemical properties, which prevents easy recognition by sequence homology. This gene encodes a 39S subunit protein. This gene and the gene for a semaphorin class 4 protein (SEMA4G) overlap at map location 10q24.31 and are transcribed in opposite directions. Sequence analysis identified multiple transcript variants encoding at least four different protein isoforms.
TRUSTED_SUSTAINABILITY

Specifications

Antigen MRPL43
Applications ELISA, Western Blot, Immunocytochemistry
Classification Polyclonal
Concentration 1.18 mg/mL
Conjugate Unconjugated
Formulation PBS with 50% glycerol and 0.01% thimerosal; pH 7.3
Gene MRPL43
Gene Accession No. Q8N983, Q99N89
Gene Alias 39S ribosomal protein L43, mitochondrial; 4930442D21Rik; bMRP36a; L43mt; Mitochondrial large ribosomal subunit protein mL43; Mitochondrial ribosomal protein bMRP36a; mitochondrial ribosomal protein L43; MRPL43; MRP-L43; MRPL43 protein; putative; retina-specific 15.7 kDa protein
Gene Symbols MRPL43
Host Species Rabbit
Immunogen Recombinant fusion protein containing a sequence corresponding to amino acids 80-155 of human MRPL43 (NP_7897621).
Purification Method Affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 309440, 84545, 94067
Target Species Human, Mouse, Rat
Content And Storage -20°C, Avoid Freeze/Thaw Cycles
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.