Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
MRPL44 Antibody, Novus Biologicals™

Rabbit Polyclonal Antibody
$382.00 - $610.00
Specifications
Antigen | MRPL44 |
---|---|
Applications | Western Blot, Immunohistochemistry, Immunohistochemistry (Paraffin) |
Classification | Polyclonal |
Conjugate | Unconjugated |
Host Species | Rabbit |
Description
MRPL44 Polyclonal specifically detects MRPL44 in Human samples. It is validated for Western Blot, Immunohistochemistry, Immunohistochemistry-Paraffin.Specifications
MRPL44 | |
Polyclonal | |
Rabbit | |
Human | |
Q9H9J2 | |
65080 | |
This antibody was developed against a recombinant protein corresponding to amino acids: SGLVRLLQQGHRCLLAPVAPKLVPPVRGVKKGFRAAFRFQKELERQRLLRCPPPPVRRSEKPNWDYHAEIQ | |
Primary | |
Specificity of antibody verified on a Protein Array containing target protein plus 383 other non-specific proteins. |
Western Blot, Immunohistochemistry, Immunohistochemistry (Paraffin) | |
Unconjugated | |
RUO | |
PBS (pH 7.2) and 40% Glycerol with 0.02% Sodium Azide | |
39S ribosomal protein L44, mitochondrial, EC 3.1.26, EC 3.1.26.-, FLJ12701, FLJ13990, FLJ37688, L44MT, mitochondrial ribosomal protein L44, MRP-L44 | |
MRPL44 | |
IgG | |
Affinity Purified |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title