Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Invitrogen™ MRPL44 Polyclonal Antibody

Rabbit Polyclonal Antibody
Supplier: Invitrogen™ PA558271
Description
Immunogen sequence: SGLVRLLQQG HRCLLAPVAP KLVPPVRGVK KGFRAAFRFQ KELERQRLLR CPPPPVRRSE KPNWDYHAEI Q Highest antigen sequence identity to the following orthologs: Mouse - 85%, Rat - 87%.
Mammalian mitochondrial ribosomal proteins are encoded by nuclear genes and help in protein synthesis within the mitochondrion. Mitochondrial ribosomes (mitoribosomes) consist of a small 28S subunit and a large 39S subunit. They have an estimated 75% protein to rRNA composition compared to prokaryotic ribosomes, where this ratio is reversed. Another difference between mammalian mitoribosomes and prokaryotic ribosomes is that the latter contain a 5S rRNA. Among different species, the proteins comprising the mitoribosome differ greatly in sequence, and sometimes in biochemical properties, which prevents easy recognition by sequence homology. This gene encodes a 39S subunit protein.
Specifications
| MRPL44 | |
| Polyclonal | |
| Unconjugated | |
| MRPL44 | |
| 1810030E18Rik; 39S ribosomal protein L44, mitochondrial; 5730593H20Rik; AI463302; COXPD16; l44; L44mt; Mitochondrial large ribosomal subunit protein mL44; mitochondrial ribosomal protein L44; MRPL44; MRP-L44; RM44 | |
| Rabbit | |
| Antigen affinity chromatography | |
| RUO | |
| 65080 | |
| Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
| Liquid |
| Immunohistochemistry (Paraffin), Western Blot | |
| 0.1 mg/mL | |
| PBS with 40% glycerol and 0.02% sodium azide; pH 7.2 | |
| Q9H9J2 | |
| MRPL44 | |
| Recombinant protein corresponding to Human MRPL44. Recombinant protein control fragment (Product #RP-102814). | |
| 100 μL | |
| Primary | |
| Human | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction