Learn More
Invitrogen™ MRPL45 Polyclonal Antibody

Description
Immunogen sequence: MQHARKAGLV IPPEKSDRSI HLACTAGIFD AYVPPEGDAR ISSLSKEGLI ERTERMKKTM ASQVSIRRIK DYDANFKIKD FPEKAKDIFI EAHLCLNNSD HDRLHTLVTE HCFPDMTWDI KYKTVRWSFV ESLEPSHVVQ VRCSSMMNQG NVYGQITVRM HTRQTLAIYD RFGRLMYGQE DVPKDVLEYV VFEKQLTNPY GSWRMHTKIV PPWAPPKQPI LKTVMIPGPQ LKPEEEYEEA QGEAQKPQLA; Positive Samples: U-87MG, OVCAR3, 293T, Mouse brain, Mouse heart, Mouse kidney, Mouse liver, Mouse spleen, Rat brain; Cellular Location: Mitochondrion.
Specifications
Specifications
| Antigen | MRPL45 |
| Applications | ELISA, Immunohistochemistry (Paraffin), Western Blot, Immunocytochemistry |
| Classification | Polyclonal |
| Concentration | 0.74 mg/mL |
| Conjugate | Unconjugated |
| Formulation | PBS with 50% glycerol and 0.01% thimerosal; pH 7.3 |
| Gene | Mrpl45 |
| Gene Accession No. | Q9BRJ2, Q9D0Q7 |
| Gene Alias | 2600005P05Rik; 39S ribosomal protein L45, mitochondrial; AU043542; L45mt; MGC11321; mitochondrial ribosomal protein L45; MRPL45; MRP-L45; R75241; Unknown (protein for MGC:127097); zgc:56480 |
| Gene Symbols | Mrpl45 |
| Show More |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.