Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ MRPL46 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA590459
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA590459 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA590459 Supplier Invitrogen™ Supplier No. PA590459
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Immunogen sequence: MAAPVRRTLL GVAGGWRRFE RLWAGSLSSR SLALAAAPSS NGSPWRLLGA LCLQRPPVVS KPLTPLQEEM ASLLQQIEIE RSLYSDHELR ALDENQRLAK KKADLHDEED EQDILLAQDL EDMWEQKFLQ FKLGARITEA DEKNDRTSLN RKLDRNLVLL VREKFGDQDV WILPQAEWQP GETLRGTAER TLATLSENNM EAKFLGNAPC GHYTFKFPQA MRTESNLGAK VFFFKALLLT GDFSQAGNKG HHVWVTKDEL GDYLKPKYLA QVRRFVSDL; Positive Samples: Mouse brain; Cellular Location: Mitochondrion.

Mammalian mitochondrial ribosomal proteins are encoded by nuclear genes and help in protein synthesis within the mitochondrion. Mitochondrial ribosomes (mitoribosomes) consist of a small 28S subunit and a large 39S subunit. They have an estimated 75% protein to rRNA composition compared to prokaryotic ribosomes, where this ratio is reversed. Another difference between mammalian mitoribosomes and prokaryotic ribosomes is that the latter contain a 5S rRNA. Among different species, the proteins comprising the mitoribosome differ greatly in sequence, and sometimes in biochemical properties, which prevents easy recognition by sequence homology. This gene encodes a 39S subunit protein.
TRUSTED_SUSTAINABILITY

Specifications

Antigen MRPL46
Applications ELISA, Immunohistochemistry (Paraffin), Western Blot
Classification Polyclonal
Concentration 0.57 mg/mL
Conjugate Unconjugated
Formulation PBS with 50% glycerol and 0.01% thimerosal; pH 7.3
Gene MRPL46
Gene Accession No. Q9EQI8, Q9H2W6
Gene Alias 3110052F15Rik; 39S ribosomal protein L46, mitochondrial; C15orf4; C80107; L46mt; LIECG2; Mitochondrial large ribosomal subunit protein mL46; mitochondrial ribosomal protein L46; Mrpl46; MRP-L46; P2ECSL
Gene Symbols MRPL46
Host Species Rabbit
Immunogen Recombinant protein (or fragment).
Purification Method Affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 26589, 67308
Target Species Human, Mouse
Content And Storage -20°C, Avoid Freeze/Thaw Cycles
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.