Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Invitrogen™ MRPL47 Polyclonal Antibody
Rabbit Polyclonal Antibody
Supplier: Invitrogen™ PA5101365
Description
Antibody detects endogenous levels of total MRPL47.
Mammalian mitochondrial ribosomal proteins are encoded by nuclear genes and help in protein synthesis within the mitochondrion. Mitochondrial ribosomes (mitoribosomes) consist of a small 28S subunit and a large 39S subunit. They have an estimated 75% protein to rRNA composition compared to prokaryotic ribosomes, where this ratio is reversed. Another difference between mammalian mitoribosomes and prokaryotic ribosomes is that the latter contain a 5S rRNA. Among different species, the proteins comprising the mitoribosome differ greatly in sequence, and sometimes in biochemical properties, which prevents easy recognition by sequence homology. This gene encodes a 39S subunit protein. This gene is immediately adjacent to the gene for BAF complex 53 kDa subunit protein a (BAF53a), in a tail-to-tail orientation. Two transcript variants encoding different protein isoforms have been identified.
Specifications
MRPL47 | |
Polyclonal | |
Unconjugated | |
MRPL47 | |
39S ribosomal protein L47, mitochondrial; 4833424P18Rik; CGI-204; ENSMUSG00000051761; Gm9859; L47mt; metanephros-derived tubulogenic factor; Mitochondrial large ribosomal subunit protein uL29m; mitochondrial ribosomal protein L47; Mrpl47; MRP-L47; MTF/L47; nasopharyngeal carcinoma metastasis-related 1; nasopharyngeal carcinoma metastasis-related protein 1; NCM1 | |
Rabbit | |
Affinity Chromatography | |
RUO | |
57129, 74600 | |
-20°C | |
Liquid |
Western Blot, Immunocytochemistry | |
1 mg/mL | |
PBS with 50% glycerol and 0.02% sodium azide; pH 7.4 | |
Q8K2Y7, Q9HD33 | |
MRPL47 | |
A synthesized peptide derived from human MRPL47, corresponding to a region within C-terminal amino acids. | |
100 μL | |
Primary | |
Human, Mouse | |
Antibody | |
IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction