Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Invitrogen™ MRPL47 Polyclonal Antibody
Rabbit Polyclonal Antibody
Supplier: Invitrogen™ PA558002
Description
Immunogen sequence: LKERNMLLTL EQEAKRQRLP MPSPERLDKV VDSMDALDKV VQEREDALRL LQTGQERARP GAWRRDIFGR IIWHKFKQWV I Highest antigen sequence identity to the following orthologs: Mouse - 91%, Rat - 93%.
Mammalian mitochondrial ribosomal proteins are encoded by nuclear genes and help in protein synthesis within the mitochondrion. Mitochondrial ribosomes (mitoribosomes) consist of a small 28S subunit and a large 39S subunit. They have an estimated 75% protein to rRNA composition compared to prokaryotic ribosomes, where this ratio is reversed. Another difference between mammalian mitoribosomes and prokaryotic ribosomes is that the latter contain a 5S rRNA. Among different species, the proteins comprising the mitoribosome differ greatly in sequence, and sometimes in biochemical properties, which prevents easy recognition by sequence homology. This gene encodes a 39S subunit protein. This gene is immediately adjacent to the gene for BAF complex 53 kDa subunit protein a (BAF53a), in a tail-to-tail orientation. Two transcript variants encoding different protein isoforms have been identified.
Specifications
MRPL47 | |
Polyclonal | |
Unconjugated | |
MRPL47 | |
39S ribosomal protein L47, mitochondrial; 4833424P18Rik; CGI-204; ENSMUSG00000051761; Gm9859; L47mt; metanephros-derived tubulogenic factor; Mitochondrial large ribosomal subunit protein uL29m; mitochondrial ribosomal protein L47; Mrpl47; MRP-L47; MTF/L47; nasopharyngeal carcinoma metastasis-related 1; nasopharyngeal carcinoma metastasis-related protein 1; NCM1 | |
Rabbit | |
Antigen Affinity Chromatography | |
RUO | |
57129 | |
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
Liquid |
Immunohistochemistry (Paraffin) | |
0.6 mg/mL | |
PBS with 40% glycerol and 0.02% sodium azide; pH 7.2 | |
Q9HD33 | |
MRPL47 | |
Recombinant protein corresponding to Human MRPL47. Recombinant protein control fragment (Product #RP-96340). | |
100 μL | |
Primary | |
Human | |
Antibody | |
IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction