Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ MRPL48 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA559117
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA559117 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA559117 Supplier Invitrogen™ Supplier No. PA559117
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Immunogen sequence: KPIYSVGGIL LSISRPYKTK PTHGIGKYKH LIKAEEPKKK KGKVEVRAIN LGTDYEYGVL NIHLTAYDMT LAESYAQYVH NLCNS Highest antigen sequence identity to the following orthologs: Mouse - 78%, Rat - 79%.

Mammalian mitochondrial ribosomal proteins are encoded by nuclear genes and help in protein synthesis within the mitochondrion. Mitochondrial ribosomes (mitoribosomes) consist of a small 28S subunit and a large 39S subunit. They have an estimated 75% protein to rRNA composition compared to prokaryotic ribosomes, where this ratio is reversed. Another difference between mammalian mitoribosomes and prokaryotic ribosomes is that the latter contain a 5S rRNA. Among different species, the proteins comprising the mitoribosome differ greatly in sequence, and sometimes in biochemical properties, which prevents easy recognition by sequence homology. This gene encodes a 39S subunit protein. A pseudogene corresponding to this gene is found on chromosome 6p.
TRUSTED_SUSTAINABILITY

Specifications

Antigen MRPL48
Applications Immunohistochemistry (Paraffin)
Classification Polyclonal
Concentration Lot-Specific
Conjugate Unconjugated
Formulation PBS with 40% glycerol and 0.02% sodium azide; pH 7.2
Gene MRPL48
Gene Accession No. Q96GC5
Gene Alias 1810030E20Rik; 2610028L11Rik; 39S ribosomal protein L48, mitochondrial; CGI-118; D4Ertd786e; HSPC290; L48mt; MGC13323; mitochondrial 39S ribosomal protein L48-like protein; Mitochondrial large ribosomal subunit protein mL48; mitochondrial ribosomal protein L48; Mrpl48; MRP-L48; Unknown (protein for MGC:133957)
Gene Symbols MRPL48
Host Species Rabbit
Immunogen Recombinant protein corresponding to Human MRPL48. Recombinant protein control fragment (Product #RP-97495).
Purification Method Antigen affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 51642
Target Species Human
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.