Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

MRPL49 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA545058
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA545058 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA545058 Supplier Thermo Scientific Supplier No. PA545058
Add to Cart
Add to Cart

MRPL49 Polyclonal Antibody

MRPL49 Polyclonal Antibody for Western Blot

TRUSTED_SUSTAINABILITY

Specifications

Antigen MRPL49
Applications Western Blot
Classification Polyclonal
Concentration 0.5 mg/mL
Conjugate Unconjugated
Formulation PBS with 2% sucrose and 0.09% sodium azide
Gene MRPL49
Gene Accession No. Q13405
Gene Alias 39S ribosomal protein L49, mitochondrial; 4833419D18Rik; Aa2-277; C11orf4; L49mt; MGC10656; Mitochondrial large ribosomal subunit protein mL49; mitochondrial ribosomal protein L49; Mrpl49; MRP-L49; Neighbor of FAU; neighbor of fau 1; next to FAU; NOF; NOF1; OK/SW-cl.67; Protein NOF1; Unknown (protein for MGC:142600)
Gene Symbols MRPL49
Host Species Rabbit
Immunogen Synthetic peptide directed towards the N-terminal of human MRPL49 (aa 40-89).
Purification Method Affinity Chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 740
Target Species Human
Content And Storage -20°C, Avoid Freeze/Thaw Cycles
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.