Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ MRPL49 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA5112889
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA5112889 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA5112889 Supplier Invitrogen™ Supplier No. PA5112889
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

MRPL49 Polyclonal Antibody for ELISA, IHC (P), Western Blot

Mammalian mitochondrial ribosomal proteins are encoded by nuclear genes and help in protein synthesis within the mitochondrion. Mitochondrial ribosomes (mitoribosomes) consist of a small 28S subunit and a large 39S subunit. They have an estimated 75% protein to rRNA composition compared to prokaryotic ribosomes, where this ratio is reversed. Another difference between mammalian mitoribosomes and prokaryotic ribosomes is that the latter contain a 5S rRNA. Among different species, the proteins comprising the mitoribosome differ greatly in sequence, and sometimes in biochemical properties, which prevents easy recognition by sequence homology. This gene encodes a 39S subunit protein. This gene and the gene for the HRD1 protein use in their respective 3' UTRs some of the same genomic sequence. Pseudogenes corresponding to this gene are found on chromosomes 5q and 8p.
TRUSTED_SUSTAINABILITY

Specifications

Antigen MRPL49
Applications ELISA, Immunohistochemistry (Paraffin), Western Blot
Classification Polyclonal
Concentration 0.3 mg/mL
Conjugate Unconjugated
Formulation PBS with 50% glycerol and 0.03% ProClin 300; pH 7.4
Gene MRPL49
Gene Accession No. Q13405, Q9CQ40
Gene Alias 39S ribosomal protein L49, mitochondrial; 4833419D18Rik; Aa2-277; C11orf4; L49mt; MGC10656; Mitochondrial large ribosomal subunit protein mL49; mitochondrial ribosomal protein L49; Mrpl49; MRP-L49; Neighbor of FAU; neighbor of fau 1; next to FAU; NOF; NOF1; OK/SW-cl.67; Protein NOF1; Unknown (protein for MGC:142600)
Gene Symbols MRPL49
Host Species Rabbit
Immunogen Recombinant Human 39S ribosomal protein L49, mitochondrial protein (1-166AA).
Purification Method Antigen affinity chromatography
Quantity 100 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 18120, 740
Target Species Human, Mouse
Content And Storage -20°C or -80°C if preferred
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.