Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
MRPL52 Antibody, Novus Biologicals™

Rabbit Polyclonal Antibody
Supplier: Novus Biologicals NBP186094
Description
MRPL52 Polyclonal specifically detects MRPL52 in Human, Mouse, Rat samples. It is validated for Western Blot, Immunohistochemistry, Immunocytochemistry/Immunofluorescence, Immunohistochemistry-Paraffin.Specifications
MRPL52 | |
Polyclonal | |
Western Blot 0.4 ug/ml, Immunohistochemistry 1:10-1:500, Immunocytochemistry/Immunofluorescence 1 - 4 ug/ml, Immunohistochemistry-Paraffin 1:50-1:200 | |
L52mt, mitochondrial ribosomal protein L52, MRP-L52,39S ribosomal protein L52, mitochondrial | |
Rabbit | |
Affinity Purified | |
RUO | |
122704 | |
Human, Mouse, Rat | |
IgG |
Western Blot, Immunohistochemistry, Immunocytochemistry, Immunofluorescence, Immunohistochemistry (Paraffin) | |
Unconjugated | |
PBS (pH 7.2) and 40% Glycerol with 0.02% Sodium Azide | |
MRPL52 | |
This antibody was developed against Recombinant Protein corresponding to amino acids:QWRLQQGLAANPSGYGPLTELPDWSYADGRPAPPMKGQLRRKAERETFARRVVLLSQEMDAGLQAWQLRQQKLQEEQRKQENALKPKGASLKSPLPSQ | |
0.1 mL | |
Primary | |
Specificity of human MRPL52 antibody verified on a Protein Array containing target protein plus 383 other non-specific proteins. | |
Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles. |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction