Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ MRPL54 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA590148
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA590148 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA590148 Supplier Invitrogen™ Supplier No. PA590148
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Immunogen sequence: MATKRLFGAT RTWAGWGAWE LLNPATSGRL LARDYAKKPV MKGAKSGKGA VTSEALKDPD VCTDPVQLTT YAMGVNIYKE GQDVPLKPDA EYPEWLFEMN LGPPKTLEEL DPESREYWRR LRKQNIWRHN RLSKNKRL; Positive Samples: A431, HT-29, LO2; Cellular Location: Mitochondrion.

Mammalian mitochondrial ribosomal proteins are encoded by nuclear genes and help in protein synthesis within the mitochondrion. Mitochondrial ribosomes (mitoribosomes) consist of a small 28S subunit and a large 39S subunit. They have an estimated 75% protein to rRNA composition compared to prokaryotic ribosomes, where this ratio is reversed. Another difference between mammalian mitoribosomes and prokaryotic ribosomes is that the latter contain a 5S rRNA. Among different species, the proteins comprising the mitoribosome differ greatly in sequence, and sometimes in biochemical properties, which prevents easy recognition by sequence homology. This gene encodes a 39S subunit protein. [provided by RefSeq, Jul 2008].
TRUSTED_SUSTAINABILITY

Specifications

Antigen MRPL54
Applications ELISA, Immunohistochemistry (Paraffin), Western Blot
Classification Polyclonal
Concentration 0.41 mg/mL
Conjugate Unconjugated
Formulation PBS with 50% glycerol and 0.01% thimerosal; pH 7.3
Gene Mrpl54
Gene Accession No. Q6P161
Gene Alias 0610008M19Rik; 39S ribosomal protein L54, mitochondrial; D10Sut1e; L54mt; Mitochondrial large ribosomal subunit protein mL54; mitochondrial ribosomal protein L54; Mrpl54; MRP-L54
Gene Symbols Mrpl54
Host Species Rabbit
Immunogen Recombinant fusion protein containing a sequence corresponding to amino acids 1-138 of human MRPL54 (NP_7584551).
Purification Method Affinity Chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 116541
Target Species Human
Content And Storage -20°C, Avoid Freeze/Thaw Cycles
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.