Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Invitrogen™ MRPL55 Polyclonal Antibody
Rabbit Polyclonal Antibody
Supplier: Invitrogen™ PA5103531
Description
Antibody detects endogenous levels of total MRPL55.
Mammalian mitochondrial ribosomal proteins are encoded by nuclear genes and help in protein synthesis within the mitochondrion. Mitochondrial ribosomes (mitoribosomes) consist of a small 28S subunit and a large 39S subunit. They have an estimated 75% protein to rRNA composition compared to prokaryotic ribosomes, where this ratio is reversed. Another difference between mammalian mitoribosomes and prokaryotic ribosomes is that the latter contain a 5S rRNA. Among different species, the proteins comprising the mitoribosome differ greatly in sequence, and sometimes in biochemical properties, which prevents easy recognition by sequence homology. MRPL55 encodes a 39S subunit protein. Multiple transcript variants encoding two different isoforms were identified through sequence analysis.
Specifications
MRPL55 | |
Polyclonal | |
Unconjugated | |
MRPL55 | |
2810038N09Rik; 39S ribosomal protein L55, mitochondrial; AAVG5835; L55mt; L55nt; Mitochondrial large ribosomal subunit protein bL31m; Mitochondrial large ribosomal subunit protein mL55; mitochondrial ribosomal protein L55; Mrpl55; MRP-L55; PRO19675; Unknown (protein for MGC:137378); UNQ5835/PRO19675 | |
Rabbit | |
Affinity chromatography | |
RUO | |
128308, 287356, 67212 | |
-20°C | |
Liquid |
Immunohistochemistry (Paraffin), Western Blot | |
1 mg/mL | |
PBS with 50% glycerol and 0.02% sodium azide; pH 7.4 | |
Q7Z7F7, Q9CZ83 | |
MRPL55 | |
A synthesized peptide derived from human MRPL55(Accession Q7Z7F7), corresponding to amino acid residues R67-E117. | |
100 μL | |
Primary | |
Human, Mouse, Rat | |
Antibody | |
IgG |
Safety and Handling
WARNING: Cancer - www.P65Warnings.ca.gov
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction