Learn More
Invitrogen™ MRPS10 Polyclonal Antibody

Description
Immunogen sequence: TAFGAVCRRL WQGLGNFSVN TSKGNTAKNG GLLLSTNMKW VQFSNLHVDV PKDLTKPVVT ISDE Highest antigen sequence identity to the following orthologs: Mouse - 61%, Rat - 56%.
Specifications
Specifications
| Antigen | MRPS10 |
| Applications | Immunohistochemistry (Paraffin), Western Blot |
| Classification | Polyclonal |
| Concentration | 0.1 mg/mL |
| Conjugate | Unconjugated |
| Formulation | PBS with 40% glycerol and 0.02% sodium azide; pH 7.2 |
| Gene | MRPS10 |
| Gene Accession No. | P82664 |
| Gene Alias | 1110038B19Rik; 28S ribosomal protein S10, mitochondrial; Ac1179; AW743084; Liver regeneration-related protein LRRG099; mitochondrial 28S ribosomal protein S10; mitochondrial ribosomal protein S10; Mitochondrial small ribosomal subunit protein uS10m; MRPS10; MRP-S10; MSTP040; PNAS-122; Rpms10; S10mt |
| Gene Symbols | MRPS10 |
| Show More |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.