Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Invitrogen™ MRPS10 Polyclonal Antibody

Rabbit Polyclonal Antibody
Supplier: Invitrogen™ PA556066
Description
Immunogen sequence: YEMRTLYRCL ELEHLTGSTA DVYLEYIQRN LPEGVAMEVT KTQLEQLPEH IKEPIWETLS EE Highest antigen sequence identity to the following orthologs: Mouse - 89%, Rat - 87%.
Mammalian mitochondrial ribosomal proteins are encoded by nuclear genes and help in protein synthesis within the mitochondrion. Mitochondrial ribosomes (mitoribosomes) consist of a small 28S subunit and a large 39S subunit. They have an estimated 75% protein to rRNA composition compared to prokaryotic ribosomes, where this ratio is reversed. Another difference between mammalian mitoribosomes and prokaryotic ribosomes is that the latter contain a 5S rRNA. Among different species, the proteins comprising the mitoribosome differ greatly in sequence, and sometimes in biochemical properties, which prevents easy recognition by sequence homology. This gene encodes a 28S subunit protein that belongs to the ribosomal protein S10P family. Pseudogenes corresponding to this gene are found on chromosomes 1q, 3p, and 9p.
Specifications
| MRPS10 | |
| Polyclonal | |
| Unconjugated | |
| MRPS10 | |
| 1110038B19Rik; 28S ribosomal protein S10, mitochondrial; Ac1179; AW743084; Liver regeneration-related protein LRRG099; mitochondrial 28S ribosomal protein S10; mitochondrial ribosomal protein S10; Mitochondrial small ribosomal subunit protein uS10m; MRPS10; MRP-S10; MSTP040; PNAS-122; Rpms10; S10mt | |
| Rabbit | |
| Antigen Affinity Chromatography | |
| RUO | |
| 55173 | |
| Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
| Liquid |
| Immunohistochemistry (Paraffin), Western Blot | |
| 0.1 mg/mL | |
| PBS with 40% glycerol and 0.02% sodium azide; pH 7.2 | |
| P82664 | |
| MRPS10 | |
| Recombinant protein corresponding to Human MRPS10. Recombinant protein control fragment (Product #RP-94610). | |
| 100 μL | |
| Primary | |
| Human | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction