Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
MRPS11 Antibody (OTI2E9), DyLight 680, Novus Biologicals™

Mouse Monoclonal Antibody
Supplier: Novus Biologicals NBP272782FR
Description
MRPS11 Monoclonal specifically detects MRPS11 in Human samples. It is validated for Western Blot.Specifications
MRPS11 | |
Monoclonal | |
DyLight 680 | |
PBS with 0.05% Sodium Azide | |
MRPS11 | |
Full length recombinant protein of human MRPS11 (NP_073750) produced in E.coli. | |
0.1 mL | |
64963.0 | |
Store at 4°C in the dark. |
Western Blot | |
OTI2E9 | |
Western Blot | |
28S ribosomal protein S11, mitochondrial, Cervical cancer proto-oncogene 2 protein, FLJ22512, FLJ23406, HCC-2S11mt, mitochondrial ribosomal protein S11, MRP-S11, RPMS11 | |
Mouse | |
Affinity Purified | |
Primary | |
Human | |
IgG1 |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction