Learn More
Invitrogen™ MRPS11 Monoclonal Antibody (OTI3B1)

Description
MRPS11 Monoclonal Antibody for Western Blot
Specifications
Specifications
| Antigen | MRPS11 |
| Applications | Western Blot |
| Classification | Monoclonal |
| Clone | OTI3B1 |
| Concentration | 1 mg/mL |
| Conjugate | Unconjugated |
| Formulation | PBS with 1% BSA, 50% glycerol and 0.02% sodium azide; pH 7.3 |
| Gene | MRPS11 |
| Gene Accession No. | P82912 |
| Gene Alias | 0710005I03Rik; 28S ribosomal protein S11, mitochondrial; C79873; Cervical cancer proto-oncogene 2 protein; HCC2; HCC-2; mitochondrial ribosomal protein S11; Mitochondrial small ribosomal subunit protein uS11m; MRPS11; MRP-S11; RGD1559901; RPMS11; S11mt |
| Show More |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.